Universal cable lockouts, SafeLex
With 2 m cable
Catalog # BRDY145550
Supplier: Brady
Jump to:
Product Details & Documents
The integrated hasp and cable in these cable lockouts provide a simple way to lock out multiple electrical or valve energy isolation points.
- Lightweight construction for easy handling, carrying and operation
- Device comes complete with vinyl coated steel lockout cable attachment
- Allows up to six safety padlocks for a secure lockout
Documents
Specifications
- Type:With 2 m cable
- Seq:03