Orders LinkContactUsLinkComponent
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
With 3 m cable
Catalog # BRDY145549
Supplier:   Brady
Universal cable lockouts, SafeLex
Catalog # BRDY145549
Supplier:  Brady
Universal cable lockouts, SafeLex
Universal cable lockouts, SafeLex
With 3 m cable
Price
68,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
The integrated hasp and cable in these cable lockouts provide a simple way to lock out multiple electrical or valve energy isolation points.
  • Lightweight construction for easy handling, carrying and operation
  • Device comes complete with vinyl coated steel lockout cable attachment
  • Allows up to six safety padlocks for a secure lockout
Specifications
  • Type:
    With 3 m cable
  • Seq:
    05