Orders LinkContactUsLinkComponent
Push button lockout devices
Push button lockout devices
Lockout bases, large (for 30 mm push buttons)
Catalog # BRDY130824
Supplier:   Brady
Push button lockout devices
Catalog # BRDY130824
Supplier:  Brady
Push button lockout devices
Push button lockout devices
Lockout bases, large (for 30 mm push buttons)
Price
30,30 €
Log in for availability
1 * 5 ST
Product Details & Documents
These lockout devices prevent the unauthorised use of push button emergency devices.
  • With standard cover
  • Made from clear PC, with translucent red cover
  • Suitable for 16, 22 or 30 mm push buttons
Specifications
  • Type:
    Lockout bases, large (for 30 mm push buttons)
  • Seq:
    11