Push button safety covers
Push button safety cover, for use with button style NEMA 30,5 mm, incl. white legend plate, 39,87×49,02 mm
Catalog # BRDY104602
Supplier: Brady
Jump to:
Product Details & Documents
These devices made from metal and clear PC guard against unauthorised operation or accidental activation.
- Clear Lexan cover with hinged door can be kept open for easy access when not in use
- For use with button style IEC 22,5 mm or NEMA 30,5 mm
Specifications
- Type:Push button safety cover, for use with button style NEMA 30,5 mm, incl. white legend plate
- Seq:04
- W×D×H:39,87×49,02 mm
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



