Orders LinkContactUsLinkComponent
Fuse blockout devices
Fuse blockout devices
Fuse blockout device, small, for 6,3 / 7,1 / 10,3 / 14,3 mm fuses, Red
Catalog # BRDY065690
Supplier:   Brady
Fuse blockout devices
Catalog # BRDY065690
Supplier:  Brady
Fuse blockout devices
Fuse blockout devices
Fuse blockout device, small, for 6,3 / 7,1 / 10,3 / 14,3 mm fuses, Red
Price
67,10 €
Log in for availability
1 * 6 ST
Product Details & Documents
These highly visible fuse blockout devices made from PP can be used in conjunction with Brady lockout tags.
Specifications
  • Type:
    Fuse blockout device, small, for 6,3 / 7,1 / 10,3 / 14,3 mm fuses
  • Colour:
    Red
  • Seq:
    01