ContactUsLinkComponent
Cleanroom Mop UnoSicuro S PW, 40cm mop with pockets, disposable, structured surface, pre-washed, for use with pre-impregnation systems, for use in C/D up to ISO 5 cleanrooms, non-sterile, double bag of 5pcs 1 * 5 items
Catalog # 141-1454
Cleanroom Mop UnoSicuro S PW, 40cm mop with pockets, disposable, structured surface, pre-washed, for use with pre-impregnation systems, for use in C/D up to ISO 5 cleanrooms, non-sterile, double bag of 5pcs 1 * 5 items
Catalog # 141-1454