Accessories
Extension tubing and Jet-Pen™ for dispenser up to 10 ml, FEP/PCTFE, 600 mm, I.Ø 4 mm
Catalog # 613-6199
Supplier: Socorex Isba
Jump to:
Product Details & Documents
Documents
Specifications
- Description:Extension tubing and Jet-Pen™ for dispenser up to 10 ml, FEP/PCTFE, 600 mm, I.Ø 4 mm
- Seq:02