Accessories
Telescopic feed tubing, FEP, 195 - 345 mm, for dispensers up 25 ml
Catalog # SOCO1.525.355
Supplier: Socorex Isba
Jump to:
Product Details & Documents
Documents
Specifications
- Désignation:Telescopic feed tubing, FEP, 195 - 345 mm, for dispensers up 25 ml
- Seq:21
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



