pH-Fix pH Indicator Test Strips
pH-Fix, 100 test strips with colour scale, 4,0 - 7,0, Classic flat box , 68×90×15 mm, 32 g
Fournisseur: MACHEREY-NAGEL
Jump to:
Product Details & Documents
pH-Fix indicator test strips allow fast pH testing directly at the point of interest, and are suitable for untrained users as well as analytical professionals.
- No accessories needed - always ready to use
- Brilliant colour chart for precise readings
- Long handle for sufficient hand - sample - distance
- Non bleeding - no contamination of sample solutions
The indicator dyes used for pH-Fix test strips are chemically bound to the test pads to prevent bleeding and, therefore, contamination of the sample. Instructions for use according to IVD Directive 98/79/EC - The instructions for use according to IVD Directive 98/79/EC can be requested by providing the article number and LOT number at the email address [email protected].
Caution: No hazardous material.
Documents
Certificates
Search by Lot NumberDip and Read Tests.pdf
Additional Product Details
Specifications
- pH range:4,0 - 7,0
- Description:pH-Fix, 100 test strips with colour scale
- Seq:9
- Pack type:Classic flat box , 68×90×15 mm, 32 g
- Cat. No.:662-2342
- Shelf life:4 Year(s)



