ELECTROLYT 3MOL/L KCL 1 * 250 mL
Catalog # KNICZU0960
Supplier: KNICK
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Hydrochloric acid 37% Ph. Eur.
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Bioindicator, Sterikon® plus
Supplier: Merck
Screw caps for wide neck bottles
Supplier: Kautex
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
VWR®, Nitrile Gloves
Supplier: VWR Collection