ContactUsLinkComponent
Lid for crucible dia 50mm lid dia 54mm, made of porcelain, very good chemical resistance, minimal thermal expansion, high resistance to thermal shocks, maximum operating temperature: 1100C, glazed 1 * 5 items
Catalog # MARI6838050
Supplier:  MARIENFELD
Lid for crucible dia 50mm lid dia 54mm, made of porcelain, very good chemical resistance, minimal thermal expansion, high resistance to thermal shocks, maximum operating temperature: 1100C, glazed 1 * 5 items
Catalog # MARI6838050
Supplier:  MARIENFELD
Frequently Bought Together