ContactUsLinkComponent
Von Willebrand disease is a congenital bleeding disorder caused by defects in the von Willebrand factor protein (VWF). VWF is a mult imeric glycoprotein that is found in endothelial cells, plasma and platelets, and it is involved in the coagulation o 1 * 100 µG
Catalog # BWRLAP6002
Von Willebrand disease is a congenital bleeding disorder caused by defects in the von Willebrand factor protein (VWF). VWF is a mult imeric glycoprotein that is found in endothelial cells, plasma and platelets, and it is involved in the coagulation o 1 * 100 µG
Catalog # BWRLAP6002
Frequently Bought Together