Orders LinkContactUsLinkComponent
Closures
Closures
Closure, push and turn with PE foam insert, GL 18, White, PP
Catalog # RIXI8-2201-18-W
Closures
Catalog # RIXI8-2201-18-W
Closures
Closures
Closure, push and turn with PE foam insert, GL 18, White, PP
Price
9,10 €
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Thread:
    GL 18
  • Colour:
    White
  • Description:
    Closure, push and turn with PE foam insert
  • Material:
    PP
  • Seq:
    24