Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
O-ring, nitrile, ¹¹/₁₆" int. Ø, ¹³/₁₆" ext. Ø, ¹/₁₆" thick, for Mark VI and Sturman-Masters spray chambers for AA/ICP-OES
Catalog # HEWL6910006000
Supplier:   Agilent
Spray chambers and brackets
Catalog # HEWL6910006000
Supplier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
O-ring, nitrile, ¹¹/₁₆" int. Ø, ¹³/₁₆" ext. Ø, ¹/₁₆" thick, for Mark VI and Sturman-Masters spray chambers for AA/ICP-OES
Price
5,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifications
  • Description:
    O-ring, nitrile, ¹¹/₁₆" int. Ø, ¹³/₁₆" ext. Ø, ¹/₁₆" thick, for Mark VI and Sturman-Masters spray chambers for AA/ICP-OES
  • Seq:
    03