Order Entry
Belgium
Orders LinkContactUsLinkComponent
Columns, PlusPore

Columns, PlusPore

Analytical, 2,10 mm, 250 mm
Catalog # HEWLPL1913-5500
Supplier:   Agilent
Columns, PlusPore
Catalog # HEWLPL1913-5500
Supplier:  Agilent
Columns, PlusPore

Analytical, 2,10 mm, 250 mm

Price
2 370,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The PlusPore family offers a major resolution improvement over mixed medias and wider range than single pore columns.

  • Separation is maximised over four common MW ranges
  • Small particles and large pore volumes give very fast, high resolution
  • High speed chromatography offers 6 min separations for common samples

The presence of multiple pore sizes on each particle maximises the separation efficiency while still offering a wide MW range with no dislocations.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    250 mm
  • Seq:
    03