Order Entry
Belgium
Orders LinkContactUsLinkComponent
Columns, PlusPore

Columns, PlusPore

Analytical, 2,10 mm, 250 mm
Catalog # HEWLPL1913-5325
Supplier:   Agilent
Columns, PlusPore
Catalog # HEWLPL1913-5325
Supplier:  Agilent
Columns, PlusPore

Analytical, 2,10 mm, 250 mm

Price
2 680,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The PlusPore family offers a major resolution improvement over mixed medias and wider range than single pore columns.

  • Separation is maximised over four common MW ranges
  • Small particles and large pore volumes give very fast, high resolution
  • High speed chromatography offers 6 min separations for common samples

The presence of multiple pore sizes on each particle maximises the separation efficiency while still offering a wide MW range with no dislocations.

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    2,10 mm
  • Length:
    250 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.