Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
Replacement helix seal for ICP spray chamber, for any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
Catalog # HEWLG8010-80042
Supplier:   Agilent
Spray chambers and brackets
Catalog # HEWLG8010-80042
Supplier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
Replacement helix seal for ICP spray chamber, for any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
Price
136,00 €
Log in for availability
1 * 4 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifications
  • Description:
    Replacement helix seal for ICP spray chamber, for any ICP spray chamber that uses helix fitting to secure nebulizer in spray chamber side arm
  • Seq:
    08