Dextrin
500 g
Catalog # TCIAD4657-500G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Dextrin
- CAS Number:9004-53-9
- Formula:(C₆H₁₂O₆)n
- MDL Number:MFCD00081554
- Molecular Weight:180.16 g/mol
- Merck Index:14,02953
- EINECS:232-675-4
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Drying loss:max. 10.0 %
- Ignition residue (Sulfate):max. 0.5 %
- Reducing suger:max. 5.0 %
Specifications
- Pk:500 g



