Orders LinkContactUsLinkComponent
Accessories for Continuous Level Control, SymLine®
Accessories for Continuous Level Control, SymLine®
Built-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plug
Catalog # SCTV106548
Supplier:   S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Catalog # SCTV106548
Supplier:  S.C.A.T.
Accessories for Continuous Level Control, SymLine®
Accessories for Continuous Level Control, SymLine®
Built-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plug
Price
485,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
LED's and acoustic alarm warn you in time before the fill level of your collection containers reaches the critical level. With mute / snooze function and interface for controlling external devices. For installation in your media line of the fume cupboard.
  • Built-in Signalbox2 for Safety Waste Cap and other electronical level controlers
  • Touchless alarm-resetting fields for two containers
  • Two switch inputs and two switch outputs for peripheral units
  • Incl. 230 V power supply cable with open end and clamp
Suitable e.g. for equipment from Wesemann, Waldner, Bense, Hemling, Köttermann, Kewaunee, Carlo Erba, Modus Complete, Delagrave, and many more.
Specifications
  • Description:
    Built-in Signalbox2; 2-channel (EU), for Safety waste cap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp, universal plug
  • Seq:
    1