Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mobile Boxes for Hazardous Materials

Mobile Boxes for Hazardous Materials

Mobile box blue, with blue lid, 250 l, 600×600×890 mm
Catalog # CEMG7806
Supplier:   CEMO
Mobile Boxes for Hazardous Materials
Catalog # CEMG7806
Supplier:  CEMO
Mobile Boxes for Hazardous Materials
Mobile Boxes for Hazardous Materials

Mobile box blue, with blue lid, 250 l, 600×600×890 mm

Price
362,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These mobile boxes with its stable design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also approved for transport according to ADR, RID and IMDG codes.

  • Easy to handle, with coloured lids
  • Specially shaped lid enables storage in two-layers
  • Up to eight boxes can be safely stacked on a Euro pallet
  • Beading in the container wall enables emptying with a rotary forklift
Specifications
  • Capacity:
    250 l
  • Description:
    Mobile box blue, with blue lid
  • Seq:
    05
  • W×D×H:
    600×600×890 mm