Mobile Boxes for Hazardous Materials
Mobile box blue, with red lid, 250 l, 600×600×890 mm
Catalog # CEMG7805
Supplier: CEMO
Jump to:
Product Details & Documents
These mobile boxes with its stable design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also approved for transport according to ADR, RID and IMDG codes.
- Easy to handle, with coloured lids
- Specially shaped lid enables storage in two-layers
- Up to eight boxes can be safely stacked on a Euro pallet
- Beading in the container wall enables emptying with a rotary forklift
Specifications
- Capacity:250 l
- Description:Mobile box blue, with red lid
- Seq:04
- W×D×H:600×600×890 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



