Orders LinkContactUsLinkComponent
Closures
Closures
Tamper evident closure with LDPE ring and PTFE insert, 28 mm, White/white, PP
Catalog # RIXI8-1316-28-WW
Closures
Catalog # RIXI8-1316-28-WW
Closures
Closures
Tamper evident closure with LDPE ring and PTFE insert, 28 mm, White/white, PP
Price
16,80 €
Log in for availability
1 * 25 ST
Product Details & Documents
Specifications
  • Thread:
    28 mm
  • Colour:
    White/white
  • Description:
    Tamper evident closure with LDPE ring and PTFE insert
  • Material:
    PP
  • Seq:
    21