5 x PUFFER KOMPLET FÜR RT, 250 mM Tris-HCl pH 8.3, 500 mM KCl, 15 mM MgCl2, 50 mM DTT 1 * 500 ml
PEQL98-0125
5 x PUFFER KOMPLET FÜR RT, 250 mM Tris-HCl pH 8.3, 500 mM KCl, 15 mM MgCl2, 50 mM DTT 1 * 500 ml
PEQL98-0125
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection