Order Entry
Belgium
Orders LinkContactUsLinkComponent
Eyewash solution, pH neutral

Eyewash solution, pH neutral

Eyewash solution, pH neutral, 200 ml, bottle in single carton
Catalog # 118-0479
Supplier:   Plum Safety
Eyewash solution, pH neutral
Catalog # 118-0479
Supplier:  Plum Safety
Eyewash solution, pH neutral
Eyewash solution, pH neutral

Eyewash solution, pH neutral, 200 ml, bottle in single carton

Price
17,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Sterile phosphate buffer solution (4,9%) for fast and effective neutralisation of acids and alkalis.

  • Handy sized bottle, well suited for use in first aid kits and emergency bags
  • Bottle with ergonomic eye cup, dust cap and clear instructions
  • Can be used as refill for Plum wall-mounted eyewash station
  • No maintenance and cleaning of the bottles required
  • Bottles in single carton: Carton protects the bottles from external contaminations and can be opened quickly and easily at the perforation

Rinsing time: ~2 minutes per 200 ml
Shelf life: 3 years

According to DIN EN 15154-4

Certifications: According to DIN EN 15154-4.

Specifications
  • Description:
    Eyewash solution, pH neutral, 200 ml, bottle in single carton
  • Seq:
    02
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Belt bag for 200 ml eyewash bottles
    Belt bag for 200 ml eyewash bottles
    118-1704
    Log in for availability
    1 * 1 ST - 9,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    Blue wall-mountable holder for 200 ml pH neutral eyewash bottles
    118-1712
    Log in for availability
    1 * 1 ST - 5,10 €