Orders LinkContactUsLinkComponent
Microplate reader, SpectraMax® L
Microplate reader, SpectraMax® L
SpectraMax® L luminescence microplate reader
Catalog # MLDVSPECTRALCONFIG
Supplier:   Molecular Devices
Microplate reader, SpectraMax® L
Catalog # MLDVSPECTRALCONFIG
Supplier:  Molecular Devices
Microplate reader, SpectraMax® L
Microplate reader, SpectraMax® L
SpectraMax® L luminescence microplate reader
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
The SpectraMax® L microplate reader measures flash and glow luminescence assays in both 96- and 384-well plates.
  • Detect dim samples with unsurpassed sensitivity
  • Avoid saturation issues with 9 orders of dynamic range
  • Enable robust performance with autorinse injectors
  • Ensure compliance with IQ/OQ and FDA CFR 21 Part 11 compliance tools
  • Easy set-up and analysis with industry-leading SoftMax Pro software
SpectraMax® L is upgradeable to include a barcode reader, programmable injectors, and dual-channel configurations for a 2-fold increase in medium throughput. The high sensitivity, low-noise photomultiplier tubes (PMTs), and variable-volume injector configuration make this reader ideal for fast kinetic assays requiring continuous monitoring during injection and measurement.
Dual injector option includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Dual injector option (auto aperture) includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Two PMTs with automatic switching option enables wavelength-based measurements such as bioluminescence resonance energy transfer (BRET) assays and a 2-fold increase in throughput for traditional luminescence measurements. Wavelength-based measurements require purchase of appropriate filters.
Four injector option is suitable for SpectraMax® L readers with two PMTs and includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Certifications: For full details about the options available with this system please contact your local Avantor sales office.
Delivery information: System includes SpectraMax® L luminometer for reading 96- and 384- well microplates with current version of SoftMax Pro software, AutoRange for >9 orders of dynamic range, Photon Counting for high sensitivity, landscape/portrait plate reading, incubator, user manual, USB interface cable, 1 year MDCare PhD Technical Support Plan and 1 year warranty covering parts and labour.
Specifications
  • Description:
    SpectraMax® L luminescence microplate reader
  • Seq:
    01
Complementary Products
Product Information
  • Catalog #
    Description
    Serum format
    Sterility
    Pk
    Recommended working volume
    Colour
    Media Format
    Packed
    Culture area
    Sterile
    Availability
    Price
  • MLDV0200-6017
    40-plate stacker magazines (pair)
    Log in for availability
    1 * 1 ST - 0,00 €
  • MLDV0200-6016
    20-plate stacker magazines (pair)
    Log in for availability
    1 * 1 ST - 0,00 €
  • MLDV0200-6187
    50-plate stacker magazines (pair)
    Log in for availability
    1 * 1 ST - 0,00 €
  • MLDV5005331
    Loading tool for facilitating loading and unloading plates
    Log in for availability
    1 * 1 ST - 0,00 €
  • S1810-500
    Foetal bovine serum, South American origin
    Sterile filtered
    15
    Log in for availability
    1 * 500 mL - 330,00 €
  • MLDVSTAKMAX
    StakMax® microplate handling system
    Log in for availability
    1 * 1 ST - 0,00 €
  • 30028-L1
    Steady-Luc™ Firefly HTS assay kit (lyophilised)
    Log in for availability
    1 * 1 EA - 150,00 €
  • 30081-1
    Firefly and Renilla Luciferase Single Tube Assay
    Log in for availability
    1 * 1 EA - 326,00 €
  • X0930-100
    Trypsin-EDTA 10X
    11
    Log in for availability
    1 * 100 mL - 21,90 €
  • 734-2097
    200 µl
    06
    Transparent
    Log in for availability
    1 * 50 ST - 197,00 €
  • L0135-500
    Ham's F12 with L-glutamine
    500 ml
    67
    Log in for availability
    1 * 500 mL - 15,60 €
  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Log in for availability
    1 * 320 ST - 231,00 €
  • 734-2327
    1/bag
    96-well plates, flat bottom, TC-treated
    0,33 cm²
    +
    17
    Clear
    Log in for availability
    1 * 100 ST - 238,00 €
  • CORN3904BC
    20/bag
    +
    75 - 200 µl
    21
    360 µl
    Black
    Log in for availability
    1 * 100 ST - 1 160,00 €