Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tissue embedding cassettes, mega Type

Tissue embedding cassettes, mega Type

Pink
Catalog # KART2925-13
Supplier:   KARTELL
Tissue embedding cassettes, mega Type
Catalog # KART2925-13
Supplier:  KARTELL
Tissue embedding cassettes, mega Type
Tissue embedding cassettes, mega Type

Pink

Price
463,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Material: POM. Tissue embedding cassettes Mega Type for samples with big dimensions. Dimension: 28×40×13,6 mm.

Specifications
  • Colour:
    Pink