Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Xylan (du noyau de maïs)

25 g
Catalog # TCIAX0078-25G
Supplier:   TCI
Xylan (du noyau de maïs)
Catalog # TCIAX0078-25G
Supplier:  TCI
CAS Number:  9014-63-5
Xylan (du noyau de maïs)

25 g

CAS Number:  9014-63-5
Price
46,20 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:9014-63-5
  • MDL Number:MFCD00082148
  • EINECS:232-760-6
Specifications

Specification Test Results

  • Appearance:
    White to Yellow to Orange powder to crystal
  • Xylose:
    min. 75.0 % (after hydrolysis)
  • Drying loss:
    max. 15.0 %

Specifications

  • Cdt:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.