Orders LinkContactUsLinkComponent
Conductivity standards, KCL
Conductivity standards, KCL
Potassium chloride standard solution, 0,1 M, 12,88 mS/cm, 500 mL
Catalog # HACHC20C250
Supplier:   Hach
Conductivity standards, KCL
Catalog # HACHC20C250
Supplier:  Hach
CAS Number:  
Conductivity standards, KCL
Conductivity standards, KCL
Potassium chloride standard solution, 0,1 M, 12,88 mS/cm, 500 mL
CAS Number:  
Price
53,50 €
Log in for availability
1 * 500 mL
Product Details & Documents
The potassium chloride standard solutions are used for calibrating conductivity cells.
Specifications
Specifications
  • Pk:
    500 mL
  • Description:
    Potassium chloride standard solution, 0,1 M
  • Conductivity:
    12,88 mS/cm
Specifications