Handheld Digital Microscopes, Universal
Product Details & Documents
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
- High speed USB 2.0 connection
- Metal or composite housing
- Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
- Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
- Integrated polariser greatly reduces glare on reflective surfaces (some models only)
Models with Automatic Magnification Reading (AMR): Allows the included software to read the magnification setting from the microscope device, simplifying the measuring process.
Models with Extended Depth of Field (EDOF): Automatically capture and combine multiple images at different focus levels to produce a final image with greater depth of field.
Models with Extended Dynamic Range (EDR): Automatically capture and combine multiple images at different exposure levels to create an image with greater dynamic range.
Models with Flexible LED Control (FLC): Additional LED controls within the included software for adjusting LED brightness and on/off toggle by quadrant.
Delivery information: Supplied with protecting pouch, software CD (DinoCapture 2.0 (Windows), DinoXcope (Mac OS)) and calibration target.
Documents
Dino-Lite Powerful Portable Versatile Dino-Lite Software
Product Literature
Specifications
- Magnification:10 - 220×
- Model:UNIVERSAL, AM7915MZT
- Resolution:5,0 MP
- Description:Handheld microscope with exchangeable caps + polariser, Edge
- Seq:5
- Additional features:AMR/EDOF/EDR/FLC
- Illumination:8 LEDs
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)Extension pole RK-06-PL, to double the height of the RK-06A / RK-06-AE stand (required when working with Long Working Distance and Extra Long Working Distance microscopes)IDCPRK-06-PLLog in for availability1 * 1 ST - 28,60 €
Product Information
- Catalog #AvailabilityPrice
- Foot pedal, enables users to take a picture with their footFoot pedal, enables users to take a picture with their foot630-1821Log in for availability1 * 1 ST - 63,30 €
Product Information
- Catalog #AvailabilityPrice
- Sturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal armSturdy high-end stand for Dino-Lite microscopes, RK-10A, incl. HD-P1 holder and RK-10-EX horizontal arm630-2517Log in for availability1 * 1 ST - 340,00 €
Product Information
- Catalog #W×D×HAvailabilityPrice
- Tabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EXTabletop stand for handheld Dino-Lite microscopes, RK-06-A, incl. extension arm RK-10-EXIDCPRK-06A150×220×270 mmLog in for availability1 * 1 ST - 252,00 €
Product Information
- Catalog #AvailabilityPrice
- Wi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion batteryWi-Fi streamer with operating manual, power adapter and 1880 Ah Li-Ion battery630-2923Log in for availability1 * 1 ST - 329,00 €
Product Information
- Catalog #W×D×HAvailabilityPrice
- Tabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EXTabletop stand for handheld Dino-Lite microscopes, RK-06-AE, with ESD-safe coating, incl. extension arm RK-10-EXIDCPRK-06AE150×220×270 mmLog in for availability1 * 1 ST - 249,00 €