ContactUsLinkComponent
Radiation Chemical Process Indicators (CPIs), for use with Meto® Label Applicators, each Label is 25 mm (1”) with Indicator Strip wh ich Transitions from Yellow to Red, 1,000 Indicators/roll. ISO 11140-1 Class 1 Compliant 1 * 1.000 Roll
Catalog # NAMSCPI-R06
Supplier:  CROSSTEX
Radiation Chemical Process Indicators (CPIs), for use with Meto® Label Applicators, each Label is 25 mm (1”) with Indicator Strip wh ich Transitions from Yellow to Red, 1,000 Indicators/roll. ISO 11140-1 Class 1 Compliant 1 * 1.000 Roll
Catalog # NAMSCPI-R06
Supplier:  CROSSTEX
Frequently Bought Together