ContactUsLinkComponent
Spore Suspension, Bacillus pumilus Cell Line 27142, 10 mL, 109 population level per 0.1 mL, ISO 11138 and USP compliant where appli cable 1 * 10 mL
Catalog # NAMSVBP-109
Supplier:  CROSSTEX
Spore Suspension, Bacillus pumilus Cell Line 27142, 10 mL, 109 population level per 0.1 mL, ISO 11138 and USP compliant where appli cable 1 * 10 mL
Catalog # NAMSVBP-109
Supplier:  CROSSTEX
Frequently Bought Together