ContactUsLinkComponent
Spore Wire Inoculated Carriers for Monitoring Hydrogen Peroxide/Plasma Processes, Geobacillus stearothermophilus Cell Line 7953, Bra ided Steel Wires Each 40 mm in length and <2 mm in Diameter, Packaged in Bulk 100 per box, 106 population level, ISO 1 * 1 Each
Catalog # NAMSSWH-06
Supplier:  CROSSTEX
Spore Wire Inoculated Carriers for Monitoring Hydrogen Peroxide/Plasma Processes, Geobacillus stearothermophilus Cell Line 7953, Bra ided Steel Wires Each 40 mm in length and <2 mm in Diameter, Packaged in Bulk 100 per box, 106 population level, ISO 1 * 1 Each
Catalog # NAMSSWH-06
Supplier:  CROSSTEX
Frequently Bought Together