ContactUsLinkComponent
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Geobacillus stearothermophilus Cell Line 7953, Stainless Steel Discs each 6 mm in Diameter, Individually Packaged in Tyvek®/Mylar® Pouches, 100 per box, 106 p 1 * 1 Each
Catalog # NAMSSDS-06
Supplier:  CROSSTEX
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Geobacillus stearothermophilus Cell Line 7953, Stainless Steel Discs each 6 mm in Diameter, Individually Packaged in Tyvek®/Mylar® Pouches, 100 per box, 106 p 1 * 1 Each
Catalog # NAMSSDS-06
Supplier:  CROSSTEX
Frequently Bought Together