ContactUsLinkComponent
Spore Suspension, Bacillus subtilis Cell Line 6633, 10 mL, 105 population level per 0.1mL, ISO 11138 and USP compliant where applic able 1 * 10 mL
Catalog # NAMSSBS-05
Supplier:  CROSSTEX
Spore Suspension, Bacillus subtilis Cell Line 6633, 10 mL, 105 population level per 0.1mL, ISO 11138 and USP compliant where applic able 1 * 10 mL
Catalog # NAMSSBS-05
Supplier:  CROSSTEX
Frequently Bought Together