Order Entry
Belgium
Orders LinkContactUsLinkComponent
Human Recombinant IL-4

Human Recombinant IL-4

100 µG
Catalog # STMC78045
Marketsource
Human Recombinant IL-4
Catalog # STMC78045
Marketsource
CAS Number:  
Human Recombinant IL-4
Human Recombinant IL-4

100 µG

CAS Number:  
Price
633,00 €
Log in for availability
1 * 1 EA
Product Details & Documents

Human Interleukin 4 (IL-4) is important for immune responses to helminth infection as well as in allergic responses (Olpihant et al.). The IL-4 receptor consists of a heterodimer of IL-4Ra and common gamma chain. IL-4 receptor engagement leads to the activation of JAK1/3 and the recruitment of STAT6 and IRS1/2 (Nelms et al.). IL-4 drives immunoglobulin class switching in B cells (to IgE, IgG4), mast cell hyperplasia, mucus production, and the differentiation of naïve T cells into Th2 cells, which produce IL-4, IL-5, IL-10, and IL-13 (Bao et al.; Nelms et al.; Olpihant et al.). In addition to Th2 cells, IL-4 is produced by CD4+ NK T cells, γ/δ T cells, activated basophils, eosinophils, and mast cells. Human IL-4 does not cross-react with mouse cells (Park et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78045
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-4
  • Purity:
    ≥95%
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.