pH/mV/°C meter, handheld, Eutech pH 150
pH/mV/°C meter, handheld, Eutech pH 150 (Set 1)
Catalog # 662-2272
Supplier: EUTECH
Jump to:
Product Details & Documents
This portable meter features a waterproof design with IP67 rating.
- Large, three-line LCD displays pH or mV values
- Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
- Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
- Stores up to 150 measurements along with temperature
- Can be used with two AA batteries or with optional universal power adapter (not included)
- Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter can be used with pH electrodes with BNC connector.
Delivery information: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, pH buffers, storage solution and rinse solution.
Documents
eutech-150-450-series-family-brochure
Product Literature
Specifications
- Description:pH/mV/°C meter, handheld, Eutech pH 150 (Set 1)
- Seq:02
- mV range:±2000
- Compliance:IP67
- pH range:−2,00...+16,00
- Data logging capacity:150 data sets
- Power supply:Two AA batteries (included), AC adapter (optional)
- Temperature compensation:Automatic or manual from −10 to +110 °C
- mV calibration:±200
- mV resolution:0,1 (±999,9); 1 (beyond)
- pH calibration:Up to 5 points (USA & NIST); up to 6 points (DIN)
- Temperature resolution:0,1 °C
- Temperature accuracy:±0,5 °C
- Weight:0,5 kg
- Cat. No.:662-2272
- pH accuracy:±0,01
- pH resolution:0,01
- Model:pH 150
- W×D×H:200×83×57 mm
- Temperature range:−10,0...+110,0 °C
- mV accuracy:±0,2 +2 LSD or +0,5 % of reading; whichever is greater
- Temperature calibration:Offset in 0,1 increments, ±10 °C
- Display:Three line custom LCD



