Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH/mV/°C meter, handheld, Eutech pH 150

pH/mV/°C meter, handheld, Eutech pH 150

pH/mV/°C meter, handheld, Eutech pH 150 (meter only)
Catalog # 662-2300
Supplier:   EUTECH
pH/mV/°C meter, handheld, Eutech pH 150
Catalog # 662-2300
Supplier:  EUTECH
pH/mV/°C meter, handheld, Eutech pH 150
pH/mV/°C meter, handheld, Eutech pH 150

pH/mV/°C meter, handheld, Eutech pH 150 (meter only)

Price
470,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

This portable meter features a waterproof design with IP67 rating.

  • Large, three-line LCD displays pH or mV values
  • Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
  • Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
  • Stores up to 150 measurements along with temperature
  • Can be used with two AA batteries or with optional universal power adapter (not included)
  • Built-in stand with wall-mount option allows for easy bench top measurement or storage

The meter can be used with pH electrodes with BNC connector.

Delivery information: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, pH buffers, storage solution and rinse solution.

Documents

Specifications
  • Description:
    pH/mV/°C meter, handheld, Eutech pH 150 (meter only)
  • Seq:
    01
  • mV range:
    ±2000
  • Compliance:
    IP67
  • pH range:
    −2,00...+16,00
  • Data logging capacity:
    150 data sets
  • Power supply:
    Two AA batteries (included), AC adapter (optional)
  • Temperature compensation:
    Automatic or manual from −10 to +110 °C
  • mV calibration:
    ±200
  • mV resolution:
    0,1 (±999,9); 1 (beyond)
  • pH calibration:
    Up to 5 points (USA & NIST); up to 6 points (DIN)
  • Temperature resolution:
    0,1 °C
  • Temperature accuracy:
    ±0,5 °C
  • Weight:
    0,5 kg
  • Cat. No.:
    662-2300
  • pH accuracy:
    ±0,01
  • pH resolution:
    0,01
  • Model:
    pH 150
  • W×D×H:
    200×83×57 mm
  • Temperature range:
    −10,0...+110,0 °C
  • mV accuracy:
    ±0,2 +2 LSD or +0,5 % of reading; whichever is greater
  • Temperature calibration:
    Offset in 0,1 increments, ±10 °C
  • Display:
    Three line custom LCD