pH/mV/°C meter, handheld, Eutech pH 150
Product Details & Documents
This portable meter features a waterproof design with IP67 rating.
- Large, three-line LCD displays pH or mV values
- Grip-Clip™: Quickly and easily secures the pH electrode to a beaker or container
- Automatic buffer recognition; up to 5- or 6-point calibration; calibration due alarm
- Stores up to 150 measurements along with temperature
- Can be used with two AA batteries or with optional universal power adapter (not included)
- Built-in stand with wall-mount option allows for easy bench top measurement or storage
The meter can be used with pH electrodes with BNC connector.
Delivery information: Meter only (without electrodes) is supplied with Grip-Clip™ and two AA batteries.
Set 1 additionally includes a double junction pH electrode and ATC probe.
Set 2 additionally includes a double-junction pH electrode and ATC probe, hard carry case, pH buffers, storage solution and rinse solution.
Set 3 additionally includes a double-junction pH/ATC electrode, hard carry case, pH buffers, storage solution and rinse solution.
Documents
eutech-150-450-series-family-brochure
Product Literature
Spécifications
- Description:pH/mV/°C meter, handheld, Eutech pH 150 (Set 3)
- Seq:04
- mV range:±2000
- Compliance:IP67
- pH range:−2,00...+16,00
- Data logging capacity:150 data sets
- Power supply:Two AA batteries (included), AC adapter (optional)
- Temperature compensation:Automatic or manual from −10 to +110 °C
- mV calibration:±200
- mV resolution:0,1 (±999,9); 1 (beyond)
- pH calibration:Up to 5 points (USA & NIST); up to 6 points (DIN)
- Temperature resolution:0,1 °C
- Temperature accuracy:±0,5 °C
- Weight:0,5 kg
- Cat. No.:662-2311
- pH accuracy:±0,01
- pH resolution:0,01
- Model:pH 150
- W×D×H:200×83×57 mm
- Temperature range:−10,0...+110,0 °C
- mV accuracy:±0,2 +2 LSD or +0,5 % of reading; whichever is greater
- Temperature calibration:Offset in 0,1 increments, ±10 °C
- Display:Three line custom LCD
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



