Labels, pipe marking
GHS labels with symbols GHS06, GHS08 and GHS05, Polyester, Brown, 250×26 mm, 4 labels/card
BRDYN005163
: Brady
These pipe marking labels are made of high quality polyester and meet with GHS (Global Harmonised System) regulations for chemical substances and mixtures.
- Promotes safety and simplifies maintenance
- Arrows are pre-cut, and hence does not requires scissors additionally
- Resistant to temperatures from –40 to +120 °C
- Resistant to ultraviolet rays and conditions
The markers have a frame that contrasts with the color of the text to increase visibility.
- Colour:Brown
- Description:GHS labels with symbols GHS06, GHS08 and GHS05
- Dimensions:250×26 mm
- Material:Polyester
- Seq:18
- Packed:4 labels/card
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



