Orders LinkContactUsLinkComponent
VWR®, Storage Boxes
VWR®, Storage Boxes
120×90×50 mm
Catalog # 216-4334
Supplier:   VWR Collection
VWR®, Storage Boxes
Catalog # 216-4334
Supplier:  VWR Collection
VWR®, Storage Boxes
VWR®, Storage Boxes
120×90×50 mm
Price
8,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
These boxes are an ideal choice for laboratories due to their high transparency and excellent durability.
  • Stackable, transparent boxes
  • For storage and presentation
  • Six different sizes
  • PS, clear
Specifications
  • Seq:
    03
  • W×D×H:
    120×90×50 mm