Orders LinkContactUsLinkComponent
VWR®, Storage Boxes
VWR®, Storage Boxes
240×180×100 mm
Catalog # 216-4002
Supplier:   VWR Collection
VWR®, Storage Boxes
Catalog # 216-4002
Supplier:  VWR Collection
VWR®, Storage Boxes
VWR®, Storage Boxes
240×180×100 mm
Price
30,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
These boxes are an ideal choice for laboratories due to their high transparency and excellent durability.
  • Stackable, transparent boxes
  • For storage and presentation
  • Six different sizes
  • PS, clear
Specifications
  • Seq:
    05
  • W×D×H:
    240×180×100 mm