Cryoboxes, PP
7×7, Green, 133×133×50/75 mm
Catalog # RATI5320313
Supplier: RATIOLAB
Aller à :
Product Details & Documents
Boxes made of high quality PP.
- Autoclavable
- Temperature-resistant from –90 to +121 °C
- Numerically coded
- Very good chemical resistance
With an numerical coding and corresponding vents for draining condensed water. The boxes are adjustable in height.
Spécifications
- Compartments:7×7
- W×D×H ext.:133×133×50/75 mm
- Colour:Green
- Seq:01