ContactUsLinkComponent
Anti-human IgG Fc Capture Biosensors - Twenty trays of 96 biosensors for the immobilization of human IgG or other proteins containi ng the human Fc region for kinetic analysis. 1 * 20 items
Catalog # PALF18-5064
Supplier:  PALL FORTEBIO
Anti-human IgG Fc Capture Biosensors - Twenty trays of 96 biosensors for the immobilization of human IgG or other proteins containi ng the human Fc region for kinetic analysis. 1 * 20 items
Catalog # PALF18-5064
Supplier:  PALL FORTEBIO
Frequently Bought Together