ContactUsLinkComponent
Anti-mouse IgG Fc Capture Biosensors - Twenty trays of 96 biosensors for the immobilization of mouse IgG or other proteins containi ng the mouse Fc region for kinetic analysis. 1 * 20 items
Catalog # PALF18-5090
Supplier:  PALL FORTEBIO
Anti-mouse IgG Fc Capture Biosensors - Twenty trays of 96 biosensors for the immobilization of mouse IgG or other proteins containi ng the mouse Fc region for kinetic analysis. 1 * 20 items
Catalog # PALF18-5090
Supplier:  PALL FORTEBIO
Frequently Bought Together