ContactUsLinkComponent
Anti-Human Fab-CH1 2nd Generation (FAB2G) Biosensors - Twenty tray of 96 biosensors coated with anti-human Fab-CH1 affinity ligand for quantitation and kinetic analysis of Fab fragments and full-length IgG. null
Catalog # PALF18-5127
Supplier:  PALL FORTEBIO
Anti-Human Fab-CH1 2nd Generation (FAB2G) Biosensors - Twenty tray of 96 biosensors coated with anti-human Fab-CH1 affinity ligand for quantitation and kinetic analysis of Fab fragments and full-length IgG. null
Catalog # PALF18-5127
Supplier:  PALL FORTEBIO
Frequently Bought Together