VWR®, Camera Adapters
C-mount adapter 0,45× for INV100/100-FL
Catalog # 630-2687
Supplier: VWR Collection
Jump to:
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Specifications
- Description:C-mount adapter 0,45× for INV100/100-FL
- Seq:03