ContactUsLinkComponent
This product is prepared by labeling high quality goat anti-rabbit IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 1 mL
  20465-1ML
 :  Biotium
This product is prepared by labeling high quality goat anti-rabbit IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 1 mL
  20465-1ML
 :  Biotium
Frequently Bought Together