ContactUsLinkComponent
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with alkaline phosphatase (AP). To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, hum 1 * 1 mL
Catalog # 20467-1ML
Supplier:  Biotium
This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with alkaline phosphatase (AP). To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, hum 1 * 1 mL
Catalog # 20467-1ML
Supplier:  Biotium
Frequently Bought Together