ContactUsLinkComponent
This product is prepared by single labeling (DOL=1) highly cross-adsorbed donkey anti-mouse IgG (H+L) with CF™535ST for (d)-STORM su per-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat 1 * 500 µl
Catalog # 20823-500UL
Supplier:  Biotium
This product is prepared by single labeling (DOL=1) highly cross-adsorbed donkey anti-mouse IgG (H+L) with CF™535ST for (d)-STORM su per-resolution microscopy. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat 1 * 500 µl
Catalog # 20823-500UL
Supplier:  Biotium
Frequently Bought Together