ContactUsLinkComponent
Cleanroom label, removable, 75x26mm, White Dimension : 75 x 26mm Gap between labels : 3mm Material type : 2074 (TMPP/RM/SRL) Compatible with thermal transfer, laser printing, and pen Acrylic-based adhesive, removable without residue Liner : silicone- 1 * 1 Roll
Catalog # BSNCLVWRG20747526
Supplier:  Basan
Cleanroom label, removable, 75x26mm, White Dimension : 75 x 26mm Gap between labels : 3mm Material type : 2074 (TMPP/RM/SRL) Compatible with thermal transfer, laser printing, and pen Acrylic-based adhesive, removable without residue Liner : silicone- 1 * 1 Roll
Catalog # BSNCLVWRG20747526
Supplier:  Basan
Frequently Bought Together